Learn More
MP Biomedicals™ SurePRIME DNA Polymerase
For Hot-Start PCR
Supplier: MP Biomedicals™ 11EPHSP525
Description
- Chemically modified Taq DNA polymerase
- Low background
- Highly specific amplification
Specifications
5U/μL | |
10X Incubation Buffer without Mg, 25mM MgCl2 | |
5 x 250U | |
Hot-Start PCR |