Learn More
Thermo Scientific™ SureQuant™ RAS Isoform Absolute Quantitation Module
Thermo Scientific™ SureQuant™ RAS Isoform Absolute Quantitation Module provides a complete quantitation solution for the analysis of key RAS expression using mass spectrometry (MS).
Manufacturer:  Thermo Scientific™ A40098
Catalog No. PIA40098
Add to cart
Description
Description
  • Complete - contains validated AQUA HeavyPeptide and LightPeptide mixtures, diluent, matrix, and system suitability standard including optimized acquisition and analysis methods
  • Validated - peptides rigorously tested for successful quantitation of each target peptide
  • Multiplex - able to quantitate NRAS, KRAS, and HRAS isoforms
Specifications
Specifications
Mass Spectrometry
Thermo Scientific™ Q Exactive Mass Spectrometer, Mass Spectrometer
Protein samples
Peptide
Approved for shipment on Wet or Dry Ice
Target Selection