Learn More
Thermo Scientific™ SureQuant™ TP53 Panel Relative Quantitation Module
Thermo Scientific™ SureQuant™ TP53 Panel Relative Quantitation Module provides a complete quantitation solution for the analysis of key TP53 expression and phosphorylation using mass spectrometry (MS).
Supplier: Thermo Scientific™ A40102
Description
- Complete - contains validated AQUA HeavyPeptide mix, diluent, matrix, system suitability standard including optimized acquisition and analysis methods
- Validated - peptides rigorously tested for successful quantitation of each target peptide
- Multiplex - able to quantitate native and phosphorylated TP53 protein
Specifications
Mass Spectrometry | |
Approved for shipment on Wet or Dry Ice | |
SureQuant™ | |
Mass Spectrometry | |
Peptides | |
10 Reactions |
Thermo Scientific™ Q Exactive Mass Spectrometer, Mass Spectrometer | |
Target Selection | |
1 Kit | |
Quantitation Module | |
Protein Samples |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.