Learn More
Surfactant Protein A Mouse anti-Human, Alexa Fluor™ 532, Clone: 6F10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP241344AF532
Catalog No. NP241344A32
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Surfactant Protein A Monoclonal antibody specifically detects Surfactant Protein A in Human samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Surfactant Protein A
6F10
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
COLEC4, PSAP, PSPA, PSP-A, SFTP1, SFTPA1B, SPA, SP-A, SPA1, SP-A1, surfactant protein A1B
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
SFTPA1
Native SP-A purified from bronchioaveolar lavage from proteinosis patients
0.1 mL
Primary
653509