Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surfactant Protein A Mouse anti-Human, Alexa Fluor™ 594, Clone: 6F10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241344AF594

Catalog No. NP241344A94


Add to cart

Description

Description

Surfactant Protein A Monoclonal antibody specifically detects Surfactant Protein A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Surfactant Protein A
6F10
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
SFTPA1
Native SP-A purified from bronchioaveolar lavage from proteinosis patients
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
COLEC4, PSAP, PSPA, PSP-A, SFTP1, SFTPA1B, SPA, SP-A, SPA1, SP-A1, surfactant protein A1B
Mouse
IgG1 Kappa
0.1 mL
Primary
653509.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit