Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surfactant Protein A Mouse anti-Human, Clone: 6F10, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications

$176.85

Specifications

Antigen Surfactant Protein A
Clone 6F10
Concentration 1 mg/mL
Dilution Western Blot 1:1000, ELISA 1:4000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Microarray, Protein Array
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019675
View Documents
Novus Biologicals
NBP2413440.025MG
0.025 mg
Each of 1 for $176.85
Add to cart
 
Description

Description

Surfactant Protein A Monoclonal specifically detects Surfactant Protein A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Microarray, Protein Array.
Specifications

Specifications

Surfactant Protein A
1 mg/mL
Monoclonal
Purified
RUO
10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide
653509
Native SP-A purified from bronchioaveolar lavage from proteinosis patients
Primary
Store at 4C. Do not freeze.
6F10
Western Blot 1:1000, ELISA 1:4000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Microarray, Protein Array
Unconjugated
Mouse
Human
COLEC4, PSAP, PSPA, PSP-A, SFTP1, SFTPA1B, SPA, SP-A, SPA1, SP-A1, surfactant protein A1B
SFTPA1
IgG1 κ
Protein A or G purified
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit