Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Surfactant Protein B (Precursor) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Surfactant Protein B (Precursor) Ab-1, Mouse Monoclonal Antibody.

$189.00 - $1003.00

Specifications

Antigen Surfactant Protein B (Precursor) Ab-1
Clone SPB01; same as 1-2-A5-9
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS704B
View Documents
Thermo Scientific™
MS704B
Biotinylated Ab with BSA and Azide 1mL
Each for $1,003.00
Add to cart
 
MS704B0
View Documents
Thermo Scientific™
MS704B0
Biotinylated Ab with BSA and Azide 100μL
Each for $254.00
Add to cart
 
MS704B1
View Documents
Thermo Scientific™
MS704B1
Biotinylated Ab with BSA and Azide 500μL
Each for $646.50
Add to cart
 
MS704P0
View Documents
Thermo Scientific™
MS704P0
Purified Ab with BSA and Azide 100μL
Each for $189.00
Add to cart
 
MS704P1
View Documents
Thermo Scientific™
MS704P1
Purified Ab with BSA and Azide 500μL
Each for $507.50
Add to cart
 
MS704P1ABX
View Documents
Thermo Scientific™
MS704P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $558.50
Add to cart
 
MS704PABX
View Documents
Thermo Scientific™
MS704PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $880.00
Add to cart
 
MS704R1
View Documents
Thermo Scientific™
MS704R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS704P
View Documents
Thermo Scientific™
MS704P
Purified Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS704R7
View Documents
Thermo Scientific™
MS704R7
Ready-to-use for IHC (UVLP) 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Surfactant Protein B (Precursor) Ab-1, Mouse Monoclonal Antibody.

Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventillation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.

Host Species: Mouse

Clone: SPB01; same as 1-2-A5-9

Isotype: IgG2a/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant prosurfactant protein B

Molecular Weight: 42-46kDa

Positive Control: Lung adenocarcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Surfactant Protein B (Precursor) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Murine
Cancer and Tumor Biology
Recombinant pro-surfactant protein B
SPB01; same as 1-2-A5-9
Monoclonal
RUO
Primary
IgG2a kappa
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit