Learn More
AIMS™ Surgery Board System for Small Animals Products for Science Education
For small animal surgery, procedure, and dissection platforms.
Manufacturer:  AIMS™ GSM1
 View more versions of this product
Catalog No. 14-370-212
Add to cart
Description
Description
  • Construction: High Density Polyethylene (HDPE) with excellent chemical resistance
  • Corner magnets for holding metal accessories
  • Recessed Steel Bars for use with magnetic accessories
  • Flexible LED lights - provides direct illu
Specifications
Specifications
Use for dissection of small animals
Space gel heating pad, suction cup limb restraints, magnetic base
HDPE, stainless steel (warming plate)
30.48 x 20.42m
Mouse, rats
Surgical board
2.3 X (main lens); 4.0X (inset lens)
17.78 x 10.16cm (warming plate)
100 to 240V 50 to 60Hz