Learn More
Kent Scientific SurgiSuite Multi-Functional Surgical Platform
Ideal for rodent surgery in laboratory settings. Kent Scientific SurgiSuite Multi-Functional Surgical Platform features ferro-magnetic stainless steel plate, homeothermic controls for efficient aseptic surgery.
Manufacturer:  Kent Scientific SURGISUITE
 View more versions of this product
Catalog No. 13-005-178
Add to cart
Description
Description
Platform:
  • Easy-to-clean, chemical resistant base
  • Bright LED lighting
  • Temperature monitoring
  • Channel for collecting fluids during surgical operations
  • Integrated warming by rechargeable, battery-operated far infrared warming pad
Stabilizers:
  • Multi-positional
  • Stabilizes animal or anesthesia tubing connected to nose cone or tracheal tube
Retractors:
  • Fixator jaws
  • Wire formed in loop to provide grip for a gloved hand
  • Reformed to create restraints or engage other lab apparatus that needs to be secured
Applications:
  • Surgery, tissue retraction of rodents
Specifications
Specifications
Surgical Platform
Chemical-resistant base, stainless steel surgical surface and polycarbonate overlay
5.15 x 4 in.
Surgery, Homeothermic Warming, Tissue Retraction
32.77 x 42.67 cm (Base) (12.90in x 16.80 in.), 30.23 x 20.07 cm (stainless steel area)
13.08 x 10.16 cm