Learn More
Kent Scientific SurgiSuite Multi-Functional Surgical Platform
Ideal for rodent surgery in laboratory settings. Kent Scientific SurgiSuite Multi-Functional Surgical Platform features ferro-magnetic stainless steel plate, homeothermic controls for efficient aseptic surgery.
$833.50 - $1085.00
Specifications
Product Type Surgical platform
For Use With (Application) Surgery, homeothermic warming, tissue retraction
Material Chemical-resistant base, stainless steel surgical surface and polycarbonate overlay
View More Specs
Products 2
Catalog Number Mfr. No. Size (Metric) Size (English) Dimensions (L x W) Surface Area (Metric) Heating Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Size (English) Dimensions (L x W) Surface Area (Metric) Heating Price Quantity & Availability  
13-005-178 View Documents Kent Scientific
SURGISUITE
30.23 x 20.07cm (stainless steel area) 11.90 x 7.90 in. (stainless steel area) 32.77 x 42.67cm (Base) (12.90in x 16.80 in.) 13.08 x 10.16cm Each for $833.50
Add to cart
13-005-179 View Documents Kent Scientific
SURGISUITELG
32.39 x 27.31cm (stainless steel area) 12.75 x 10.75 in. (stainless steel area) 40.01 x 45.21cm (Base) (15.75 x 17.80 in.) 22.23 x 17.15cm Each for $1,085.00
Add to cart
Description
Description
Platform:
  • Easy-to-clean, chemical resistant base
  • Bright LED lighting
  • Temperature monitoring
  • Channel for collecting fluids during surgical operations
  • Integrated warming by rechargeable, battery-operated far infrared warming pad
Stabilizers:
  • Multi-positional
  • Stabilizes animal or anesthesia tubing connected to nose cone or tracheal tube
Retractors:
  • Fixator jaws
  • Wire formed in loop to provide grip for a gloved hand
  • Reformed to create restraints or engage other lab apparatus that needs to be secured
Applications:
  • Surgery, tissue retraction of rodents
Specifications
Specifications
Surgical platform
Surgery, homeothermic warming, tissue retraction
Chemical-resistant base, stainless steel surgical surface and polycarbonate overlay