Surgitube™ Tubular Gauze

Manufacturer: Moore Medical Corp 08282

 View more versions of this product

Catalog No. 19-898-017

  Check Availability
Add to Cart

    Specifications

    Material Knitted Cotton Yarn
    Product Type Tubular Gauze
    Quantity 1 Roll
    Sterile Yes
    Color White
    For Use With (Application) Designed to protect and secure primary dressings, for hands, feet, lower legs and shoulders
    Size 3P

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit