Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Survivin Rabbit anti-Human, FITC, Clone: 1277A, Novus Biologicals™

Rabbit MonoclonalAntibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259503F

Catalog No. NBP259503F


Add to cart

Description

Description

Survivin Monoclonal antibody specifically detects Survivin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
Specifications

Specifications

Survivin
Monoclonal
FITC
PBS with 0.05% Sodium Azide
API4 survivin variant 3 alpha, Apoptosis inhibitor 4 baculoviral IAP repeat-containing protein 5, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, BIRC5, EPR-1, IAP4
Rabbit
IgG
0.1 ml
Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Cellular Markers, Core ESC Like Genes, Glia Markers, Growth and Development, Inhibitors Activators and Regulators, Mitotic Regulators, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Stem Cell Markers, Tumor Suppressors
Primary
332
Flow Cytometry, Immunocytochemistry, Immunofluorescence
1277A
Immunocytochemistry/Immunofluorescence, Flow (Intracellular)
Purified
BIRC5
Full length recombinant human Survivin [Uniprot: O15392]
Protein A or G purified from cell culture supernatant
RUO
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)