Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight Survivor™ X USB

Catalog No. 17108847
Click to view available options
Color:
Orange
Yellow

The Survivor™ X is a USB rechargeable, Class I and II, Div. 2 right angle, clip-on flashlight for firefighters. Featuring a slim profile and D-ring for accurate light orientation. Its spring-loaded clip easily attaches to belts and harnesses.

  • Class I and II, Div 2 rated, Intrinsically Safe
  • Rechargeable
  • 250 lumens and 447 meter beam distance on High
  • Choose Low mode for a longer runtime of up to 15 hours
  • High, Low, Flash modes
  • Easy operation of push button switch even with gloves on. Battery indicator in switch
  • Durable nylon construction and 2 meter impact resistance tested
  • IP67 Dust-tight and waterproof

Specifications

Light Source LED
Voltage 3.7 V
Battery Run Time 4 hrs. (High), 15 hrs. (Low)
Battery Type Lithium-ion
Beam Distance 447 meters
Candle Power 41000 Candela (High), 14000 Candela (Low)
Case Material Nylon
Certifications/Compliance Class I and II, Division 2, Groups A,B,C,D,F,G; Class III, T-Code: T4, Flashlight meets 500°F requirements of NFPA 1971-8.6 (2013), Charger meets requirements of NFPA 1901-14.1.10.2 (2016) mounted in any position, CEC Compliant
Color Yellow
Includes SL-B26 USB Battery, Battery Carrier
Lens Material Polycarbonate
Lighting Intensity 250 Lumens (High), 60 Lumens (Low)
Material Nylon
Model SURVIVOR X USB
Packaging Quantity 1
Product Line Streamlight
Warranty Limited Lifetime Warranty
Product Type Flashlight
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.