Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUSD2 Rabbit anti-Human, Clone: 1279A, R&D Systems™
Rabbit Monoclonal Antibody
$151.30 - $427.50
Specifications
Antigen | SUSD2 |
---|---|
Clone | 1279A |
Immunogen | HEK293 human embryonic kidney cell line-derived human SUSD2 Met1-Ala785 Accession # Q9UGT4 |
Host Species | Rabbit |
Primary or Secondary | Primary |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB90562SP
|
R&D Systems
MAB90562SP |
25 μg |
Each for $151.30
|
|
MAB90562100
|
R&D Systems
MAB90562100 |
100 μg |
Each for $427.50
|
|
Description
SUSD2 Monoclonal specifically detects SUSD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
SUSD2 | |
HEK293 human embryonic kidney cell line-derived human SUSD2 Met1-Ala785 Accession # Q9UGT4 | |
Primary | |
Human | |
Unconjugated | |
56241 | |
Purified | |
Western Blot, Flow Cytometry, Immunohistochemistry |
1279A | |
Rabbit | |
Monoclonal | |
IgG | |
BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
Protein A or G purified from cell culture supernatant | |
RUO |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title