Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUSD2 Rat anti-Human, Unconjugated, Clone: 19E8, MilliporeSigma™

Rat Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen SUSD2
Applications Flow Cytometry, Immunoprecipitation
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC17141MI
View Documents
MilliporeSigma
MABC1714100UL
100 μL
Each for $372.00
Add to cart
 
MABC17142MI
View Documents
MilliporeSigma
MABC171425UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

SUSD2 Monoclonal antibody specifically detects SUSD2 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications

Specifications

SUSD2
Unconjugated
Rat
Sushi domain-containing protein 2
An expression construct corresponding to the extracellular domain of human SUSD2.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Flow Cytometry, Immunoprecipitation
Purified
Human
SUSD2
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit