Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUSD5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548219

Catalog No. PA548219

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

SUSD5
Polyclonal
PBS with 0.09% sodium azide
O60279
Sushi domain-containing protein 5, SUSD5, KIAA0527
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26032
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SUSD5
Liquid
SUSD5
KLH conjugated synthetic peptide between 593-621 amino acids from the C-terminal region of human SUSD5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit