Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Suspended Painted Steel Cabinet

Fisherbrand™ Suspended Painted Steel Cabinets are durable and constructed with SEFA 8 Metal compliant load-bearing capacity.

$1191.00 - $1306.00

Specifications

Cabinet Type Suspended
Color Fisherbrand White
Depth (English) 22 in.
Height (English) 23.0625 in.
Includes Hanging Hardware
View More Specs

Products 2
Catalog Number Width (English) Price Quantity & Availability  
Catalog Number Width (English) Price Quantity & Availability  
06-000-512
View Documents
18 in. Each for $1,191.00
Add to cart
 
06-000-513
View Documents
24 in. Each for $1,306.00
Add to cart
 
Description

Description

  • Suspended Cabinets are designed to be hung from any of the Fisherbrand Table Systems.
  • Each suspended cabinet is furnished with complete hanging hardware.
  • Fisherbrand White is a urethane powder coating that provides reliable chemical and abrasion resistance.
Specifications

Specifications

Suspended
22 in.
Hanging Hardware
Cabinet
Fisherbrand White
23.0625 in.
4
2 years
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit