Learn More
Sklar InstrumentsSupplier Diversity Partner Suture Wire, Sklar, .010 in. Diameter - 180 ft. Suture Wire Veterinary Only - 30 Gauge, Stainless Steel. Sklar's Suture Wire is used for closing wounds and incisions for Veterinary care only. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 242230
Suture Wire, Sklar, .010 in. Diameter - 180 ft. Suture Wire Veterinary Only - 30 Gauge, Stainless Steel. Sklar's Suture Wire is used for closing wounds and incisions for Veterinary care only. It is also used to hold bones in place until healed from accidents. The wire comen in a veriety of diameter (gauges). This product is .010 inches (30 Gauge) and comes on a spool of 180 feet.
Catalog No. 50-143-8008
Due to product restrictions, please to purchase or view availability for this product.