Learn More
SUZ12 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259245
 View more versions of this product
Catalog No. NBP259245
Add to cart
Description
Description
SUZ12 Polyclonal antibody specifically detects SUZ12 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications
Specifications
SUZ12
Unconjugated
Serum with 0.05% Sodium Azide
ChET 9 protein, CHET9polycomb protein SUZ12, Chromatin precipitated E2F target 9 protein, JJAZ1Joined to JAZF1 protein, KIAA0160Suppressor of zeste 12 protein homolog, suppressor of zeste 12 homolog (Drosophila)
Rabbit
IgG
100 ul
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Western Blot, ChIP assay
Western Blot 11:000, Chromatin Immunoprecipitation 5 ul
Antiserum
SUZ12
The exact immunogen is propietary information.
Whole Antiserum
RUO
Primary
23512