Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SV2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104172
Description
Antibody detects endogenous levels of total SV2C.
Plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.
Specifications
SV2C | |
Polyclonal | |
Unconjugated | |
SV2C | |
4930527L09Rik; AI841971; KIAA1054; SV2C; Synaptic vesicle glycoprotein 2C; synaptic vesicle protein 2C | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22987, 29643, 75209 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q496J9, Q69ZS6, Q9Z2I6 | |
SV2C | |
A synthesized peptide derived from human SV2C(Accession Q496J9), corresponding to amino acid residues R345-I395. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction