Learn More
SV40 Large T Antigen Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ SV40 Large T Antigen Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Epredia MS1832PABX
Description
The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.Host Species: Mouse
Clone: PAb101; originally clone 412
Isotype: IgG2a1
Species Reactivity: SV40 virus.
Epitope: C-terminal
Immunogen: SV40-transformed mouse line B4
Molecular Weight: 94kDa
Positive Control: SV40-transfected cells
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
SV40 Large T Antigen Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Infectious Disease | |
Simian virus 40 (SV40) |
Immunofluorescence, Immunoprecipitation, Western Blot | |
PAb101; same as clone 412 | |
Unconjugated | |
SV40 transformed mouse line B4 | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.