Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SV40 T-antigen Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313382
This item is not returnable.
View return policy
Description
SV40 T-antigen Polyclonal antibody specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| SV40 T-antigen | |
| Polyclonal | |
| PBS, 1% BSA, 20% Glycerol (pH7) | |
| Recombinant protein encompassing a sequence within the N-terminus region of SV40 T-antigen (SV40 virus (strain 776)). The exact sequence is proprietary. | |
| 100 μg | |
| Cancer, Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction