Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SV40T-antigen Mouse anti-Virus, Janelia Fluor 646, Clone: PAB416, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259676JF646
Description
SV40 T-antigen Monoclonal specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
SV40T-antigen | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SV40 transformed mouse cells | |
0.1 mL | |
Primary | |
Virus | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
PAB416 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Mouse | |
Protein A purified | |
Cancer, Virology Bacteria and Parasites | |
Specific for N-terminal domain of SV40 large and small T-antigen | |
Store at 4°C in the dark. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title