Learn More
ATCC SW1463; Colorectal Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CCL234
Encompass
SW1463 is a cell line exhibiting epithelial morphology that was isolated from the rectum of a White, 66-year-old, female patient with colorectal adenocarcinoma. This cell line was deposited by A Leibovitz and can be used in cancer research.
Catalog No. 50-238-4911
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.