Learn More
Swift™ M3 Series Macro/Micro Field Microscopes Products for Science Education
Great for viewing live specimens and for field work applications.
$457.00
Specifications
Axis Type Coaxial Coarse and Fine
Condenser Type Built-in 0.65 N.A.
Design Type Stage clip
Diaphragm Iris
Magnification Power 4X, 10XD, 40XRD, 2X macro
View More Specs
Includes: Stage plate for viewing micro speciments, black/white contrast and clear collection containger for viewing macro specimens
Products 1
Catalog Number Mfr. No. Head Style Price Quantity & Availability  
Catalog Number Mfr. No. Head Style Price Quantity & Availability  
S98498A Products for Science Education
View Documents
Swift™
M3B
Binocular
Each for $457.00
Add to cart
Description
Description
Allow viewing of both microscopic specimens, by using the compound objectives (4X, 10XD, and 40XRD), and macroscopic specimens, by using the 1X macro objective.
  • Cordless, uses energy-efficient, rechargeable LED illumination
  • Allows up to 50 hours of use on a single 8-hour charge
  • Magnification range of 10X-400X
  • W10XD, 18mm eyepiece
  • Built-in NA 0.65 condenser
  • All-metal construction
  • Sturdy detachable tripod legs fold for storage and stability on most indoor and outdoor surfaces
  • Multiple stage positions allow for micro and macro viewing
  • Top and bottom LED illumination provides energy-efficient white light with virtually no heat
  • Ergonomic carry handle
  • Stage clips
  • Top and bottom LED illumination provides energy-efficient white light with virtually no head
  • DIN Achromatic 4X, 10XD, 40XRD objective for compound use (40X-400X magnification range)
  • DIN Achromatic 1XRD for macro use (10X magnification)
Classroom or field work applications
Warranty and Services
Three-year limited warranty
Specifications
Specifications
Coaxial Coarse and Fine
Stage clip
4X, 10XD, 40XRD, 2X macro
Viewing live specimens and for field work applications
38.8 cm
LED
3 Years
15 kg
Built-in 0.65 N.A.
Iris
WF10X/18 mm
15.25 in.
Stage plate for viewing micro specimens, black/white contrast and clear collection container for viewing macro specimens
110/220 V
6.8 lb.