Learn More
Swift™ M10T-BTW1 Series Digital Tablet Compound Microscopes Products for Science Education
Combination of compound microscope with Integrated digital tablet.
Manufacturer:  Swift™ M10TBTW1S
 View more versions of this product
Catalog No. S22236
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Micro SD card slot and mini HDMI outputs
Specifications
Specifications
LED
USB: 1280 x720/1280x 1024 preview mode, 2592x1944 (5MP) still imageWifi: 1280 x960 preview mode, 1280 x960 (1.3MP) still image
Trinocular
Semi Plan
Digital tablet compound microscope
LCD
Abbe 1.25 N.A.