Learn More
Swift™ M15-BTW1-P Digital Tablet Compound Microscope Products for Science Education
Combination of Advanced Compound level Microscope with Integrated digital tablet.
Manufacturer:  Swift™ M15TBTW1P
Catalog No. S22247
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Pre-loaded with Motic Apps to connect, view and share live images
  • Features infinity-corrected optics, 20mm  WF eyepeices
  • Built-in low-drive mechanical stage
  • Built-in camera
  • HDMI output
Specifications
Specifications
Education
Abbe condenser: 1.25 NA
Compound microscope, digital compound microscope, infinity -corrected objectives, plan objectives, LCD screen, tablet, digital imaging, microscope imaging
Halogen
USB: 1280 x720/1280x 1024 preview mode, 2592x1944 (5MP) still image Wifi: 1280 x960 preview mode, 1280 x960 (1.3MP) still image
LCD
Trinocular
Compound Microscope