Learn More
Swift™ Digital Compound Microscope Products for Science Education
Featuring a 3.0MP built-in digital camera, this Swift™ Optical Digital Compound Microscope includes Motic Images Plus software, providing you with faster image capture and annotation.
Manufacturer:  Swift™ M3602C3DGL
Catalog No. S23866
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 3.0MP built-in digital camera
  • Energy-efficient variable, cordless LED illumination provides white light with virtually no heat
  • LED light ideal for image capture and protecting live specimens
  • 4XD, 10XD and 40XRD Achromat objectives
  • Motic Images Plus software included for image capture and annotation
Specifications
Specifications
Education
3.0MP
Abbe 1.25 Spiral
W10XD, 18mm
Variable Cordless LED
M3602C-3DGL
Built-in low drive mechanical stage
4XD, 10XD, 40XRD
Digital Monocular
Microscope