Learn More
Swift™ M3F-BTW-1 Forensic Comparison Microscope with Integrated Tablet Screen Products for Science Education
To compare images in both micro and macro environments.
Manufacturer:  Swift™ M3FBTW1
Catalog No. S22239
Add to cart
Description
Description
  • Micro macro forensic comparison by adjusting stage positions and magnification options
  • Built-in WiFi signal permits user to connect to tablet or computer, using included Motic Imaging Software
  • Built-in camera for viewing images from 100% left, 100% right, side-by-side or over lapping
  • HDMI output
  • Wifi interface
  • 10 in. tablet
Specifications
Specifications
Trinocular
LED
USB: 1280 x720/1280x 1024 preview mode, 2592x1944 (5MP) still imageWifi: 1280 x960 preview mode, 1280 x960 (1.3MP) still image
Comparison microscopy, forensic microscopy studies, view live images on tablet screen
Digital
Dgital Tablet Compound Microscope
10X Macro, 40X to 400X Micro