Learn More
Fisher Science Education™ Moticam™ Digital Microscope Camera Products for Science Education
Use your own microscope to capture high-resolution still or video images with the Moticam Digital Microscope Camera.
$243.14 - $664.00
Specifications
Product Type Digital microscope camera
For Use With (Equipment) Microscopes
Compatibility (Operating System) Windows XP/Vista/7/Apple Mac OS
Sensor Types CMOS
Warranty 1 Year
View More Specs
Products 3
Catalog Number Mfr. No. Resolution Includes Required Accessories Price Quantity & Availability  
Catalog Number Mfr. No. Resolution Includes Required Accessories Price Quantity & Availability  
S02359A Products for Science Education View Documents N/A 2 MP digital signal @ 1600x1200 maximum resolution CS Ring Adaptor, Focusable Lens, 30mm and 38mm Eyepiece Adapters, USB3.0 Cable, Calibration Slide, Macro Tube, Motic Images Plus 3.0 for PC and OSX Processor: 2 Ghz or Above, USB 2.0 (minimum requirement), USB 3.0 (highly recommended), RAM: 2 GB, Hard disk: 1 GB (free space), Video card: 128 MB video memory Each for $243.14
Add to cart
S22240 Products for Science Education View Documents N/A 3 MP digital signal @ 2048x1536 maximum resolution CS Ring Adaptor, Focusable Lens, 30mm and 38mm Eyepiece Adapters, USB3.0 Cable, Calibration Slide, Macro Tube, Motic Images Plus 3.0 for PC and OSX Processor: 2 Ghz or Above, USB 2.0 (minimum requirement), USB 3.0 (highly recommended), RAM: 2 GB, Hard disk: 1 GB (free space), Video card: MUST have dedicated 256MB video memory Each for $664.00
Add to cart
S22241 Products for Science Education View Documents Fisher Science Education™
DMOTICAM 5+
5 MP digital signal @ 2592x1944 maximum resolution CS Ring Adaptor, Focusable Lens, 30mm and 38mm Eyepiece Adapters, USB3.0 Cable, Calibration Slide, Macro Tube, Motic Images Plus 3.0 for PC and OSX Processor: 2 Ghz or Above, USB 2.0 (minimum requirement), USB 3.0 (highly recommended), RAM: 2 GB, Hard disk: 1 GB (free space), Video card: MUST have dedicated 256MB video memory This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • High-resolution camera permits you to use your own microscope to create still or moving microscope images on your PC
  • Lightweight digital camera mounts over almost any microscope eyepiece (stereo or compound) with supplied C ring adapter and microscope eyepiece adapters
  • Included Motic Images Plus software allows user to view, enhance, label, measure, print, and store the images all with one program
Specifications
Specifications
Digital microscope camera
Windows XP/Vista/7/Apple Mac OS
1 Year
Microscopes
CMOS