Learn More
Swift™ Stereo Trinocular Zoom Microscope Products for Science Education
Swift™ Optical SM105T-C Stereo Trinocular Zoom Microscope is a new addition to popular SM100 Stereo Series Microscopes, that features 10 to 30X zoom magnification (1x to 3x objectives).
Manufacturer:  Swift™ SM105TC
Catalog No. S23867
Add to cart
Description
Description
  • Zoom with camera port head
  • Zoom magnification, 10 to 30X (1x to 3x objectives)
  • Top and bottom, energy-efficient LED illumination
  • Cordless and rechargeable
  • W10X eyepieces with right eyepiece diopter focusing adjustment
  • “Slip-clutch” system protects against over-focusing
  • Locked on eyepiece and stage clips
  • Trinocular head port for easy addition of cameras, such as Moticam X (camera and  c-mount camera adapter sold separately)
  • Ideal for high school classrooms and up
Specifications
Specifications
Microscope
Education
SM105T-C
W10X, 20mm
Top and bottom, LED
Limited lifetime warranty
High school classrooms and up
1X to 3X
Trinocular