Learn More
Andwin Scientific Swing-A Way portable can opener. Heavy-duty drive gears and an extra-hard cutting wheel
SDP
Supplier:  Andwin Scientific Z103403
Encompass
Useful for opening the overpack cans in which hazardous or odoriferous compounds are shipped. Chrome-plated. Has heavy-duty drive gears and an extra-hard cutting wheel mounted on a wide-diameter permanent-alignment shaft.
Catalog No. NC9836273
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.