Tocris™ SX 011

p38 MAPK inhibitor

Manufacturer: Tocris™ 3639/10

 View more versions of this product

Catalog No. 36-391-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors SX 011
    Purity >98
    Molecular Weight 483.96
    Quantity 10mg
    Product Type SX 011

    Potent p38α inhibitor (IC50 = 9nM). Also inhibits p38β and JNK-2 (IC50 values are 90nM and 100nM respectively). Displays no significant activity at p38γ, p38δ, ERK-2 and JNK-1.

    • Chemical Name: 6-Chloro-5-[[4-[(4-fluorophenyl)methyl]-1-piperidinyl]carbonyl-N,N,1-trimethyl-α-oxo-1H-indole-3-acetamide
    • Formula: C26H27ClFN3O3
    • CAS Number: 309913-42-6
    • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit