Learn More
SYCP3 Mouse anti-Human, Clone: OTI3D6, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806547
Catalog No. 50-166-7878
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregcy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.
Specifications
Specifications
SYCP3
Monoclonal
Unconjugated
SYCP3
Lyophilized
SYCP3
Full length human recombit protein of human SYCP3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3D6
PBS with 8% trehalose and no preservative; pH 7.3
Q8IZU3
COR1, RPRGL4, SCP3, SPGF4
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
50511