Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYDE1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548776

Catalog No. PA548776

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications

Specifications

SYDE1
Polyclonal
PBS with 0.09% sodium azide
Q6ZW31
Rho GTPase-activating protein SYDE1, Synapse defective protein 1 homolog 1, Protein syd-1 homolog 1, SYDE1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
85360
Western Blot
Unconjugated
SYDE1
Liquid
SYDE1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SYDE1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit