Learn More
Invitrogen™ SYF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528706
Description
Recommended positive controls: GCIP interacting protein p29 tansfected 293T cell. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (100%), Chicken (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
SYF2 | |
Polyclonal | |
Unconjugated | |
SYF2 | |
1110018L13Rik; Cbpin; CCNDBP1 interactor; CCNDBP1-interactor; D4Bwg1551e; DKFZP564O2082; fSAP29; functional spliceosome-associated protein 29; GCIP-interacting protein p29; GCIP-interacting protein p29 (Gcipip); Gcipip; mp29; Ntc31; P29; Pre-mRNA-splicing factor SYF2; SYF2; SYF2 homolog, RNA splicing factor; SYF2 homolog, RNA splicing factor (S. cerevisiae); SYF2 pre-mRNA splicing factor; SYF2 pre-mRNA-splicing factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25949 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.71 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O95926 | |
SYF2 | |
Synthetic peptide corresponding to a region within amino acids 181 and 243 of Human GCIP interacting protein p29. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.