Learn More
Alfa Aesar™ Syk Inhibitor
Manufacturer:  Alfa Aesar™ J64281LB0
 View more versions of this product
Catalog No. AAJ64281LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
2°C to 8°C
Syk Inhibitor
622387-85-3
353.40
Light Sensitive
Dark orange
Solid
A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor
C18H15N3O3S
Spleen Tyrosine Kinase Inhibitor
Soluble in DMSO