Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Syk Signaling Protein Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results Epredia™ Syk Signaling Protein Ab-1, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS322R1

Catalog No. MS322R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Syk, a protein tyrosine kinase (PTK) is expressed in immature T cells, all B cells, basophils, hepatocytes, platelets, and erythrocytes. It functions in some T cell receptor signalling processes in addition to B cell receptor and integrin-mediated signal transduction processes. syk-/syk- mice exhibit defective phagocytosis by macrophages.

Host Species: Mouse

Clone: 4D10.1

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with pig or mouse. Others not known.

Epitope: aa 313-339

Immunogen: Synthetic peptide corresponding to aa 313-339 of human Syk protein

Molecular Weight: 72kDa

Positive Control: Raji, K562, HL-60, U937 cells or lymphoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • in vitro Kinase Assay (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Syk Signaling Protein Ab-1
Monoclonal
Unconjugated
Mouse
1 mL
Signal Transduction
Human
IgG2a κ
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
4D10.1
Ready-to-use for IHC (UVLP)
Synthetic peptide corresponding to aa 313-339 of human Syk protein
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit