Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Syk Signaling Protein Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results Thermo Scientific™ Syk Signaling Protein Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen Syk Signaling Protein Ab-1
Clone 4D10.1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS322R1
View Documents
Thermo Scientific™
MS322R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Provide accurate, reproducible results Thermo Scientific™ Syk Signaling Protein Ab-1, Mouse Monoclonal Antibody.

Syk, a protein tyrosine kinase (PTK) is expressed in immature T cells, all B cells, basophils, hepatocytes, platelets, and erythrocytes. It functions in some T cell receptor signalling processes in addition to B cell receptor and integrin-mediated signal transduction processes. syk-/syk- mice exhibit defective phagocytosis by macrophages.

Host Species: Mouse

Clone: 4D10.1

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with pig or mouse. Others not known.

Epitope: aa 313-339

Immunogen: Synthetic peptide corresponding to aa 313-339 of human Syk protein

Molecular Weight: 72kDa

Positive Control: Raji, K562, HL-60, U937 cells or lymphoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • in vitro Kinase Assay (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Syk Signaling Protein Ab-1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Signal Transduction
Synthetic peptide corresponding to aa 313-339 of human Syk protein
4D10.1
Monoclonal
RUO
Primary
IgG2a, κ
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit