Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYM 2206, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

173952-44-8
C20H22N4O3
366.421
OFUDZKKOKPGXOH-UHFFFAOYSA-N
(±)-4-(4-Aminophenyl)-1, 2-dihydro-1-methyl-2-propylcarbamoyl-6, 7-methylenedioxyphthalazine
5039877

Non-competitive AMPA antagonist

Novel, potent, non-competitive AMPA receptor antagonist (IC50 = 2.8 μM ). Acts allosterically at the same regulatory site as GYKI 52466 and 53655 and other benzodiazepines but does not bind to the central diazepine binding site. Selective for AMPA relative to kainate receptor sub-types. Similar potency to GYKI 53655. Anticonvulsant in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES CCCNC(=O)N1C(C2=CC3=C(C=C2C(=N1)C4=CC=C(C=C4)N)OCO3)C
IUPAC Name 8-(4-aminophenyl)-5-methyl-N-propyl-5H-[1,3]dioxolo[4,5-g]phthalazine-6-carboxamide
Formula Weight 366.42
Chemical Name or Material SYM 2206
Grade >99%

Recommended Storage : Store at Room Temperature