Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sym-Dimethylhydrazine Dihydrochloride 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
306-37-6
C2H10Cl2N2
133.016
MFCD00012522
WLDRFJDGKMTPPV-UHFFFAOYSA-N
1,2-dimethylhydrazine dihydrochloride,n,n'-dimethylhydrazine dihydrochloride,1,2-dimethylhydrazinium dichloride,1,2-dimethylhydrazine 2hcl,sym-dimethylhydrazine dihydrochloride,hydrazine, 1,2-dimethyl-, dihydrochloride,dimethylhydrazinium dichloride,ccris 260,1,2-dimethylhydrazine.2hcl,hydrazine, 1,2-dimethyl-, hydrochloride 1:2
9380
CHEBI:73759
1,2-dimethylhydrazine;dihydrochloride
CNNC.Cl.Cl
306-37-6
C2H10Cl2N2
133.016
MFCD00012522
WLDRFJDGKMTPPV-UHFFFAOYSA-N
1,2-dimethylhydrazine dihydrochloride,n,n'-dimethylhydrazine dihydrochloride,1,2-dimethylhydrazinium dichloride,1,2-dimethylhydrazine 2hcl,sym-dimethylhydrazine dihydrochloride,hydrazine, 1,2-dimethyl-, dihydrochloride,dimethylhydrazinium dichloride,ccris 260,1,2-dimethylhydrazine.2hcl,hydrazine, 1,2-dimethyl-, hydrochloride 1:2
9380
CHEBI:73759
1,2-dimethylhydrazine;dihydrochloride
CNNC.Cl.Cl
Quantity 10g
Infrared Spectrum Authentic
Melting Point 166°C to 171°C (decomposition)
Color White to Yellow
Packaging Glass bottle
Assay Percent Range 99+%
Linear Formula CH3NHNHCH3·2HCl
Beilstein 04, 547
Merck Index 10, 3243
Formula Weight 133.02
Physical Form Crystalline Flakes or Powder
Percent Purity ≥99.0%
Chemical Name or Material sym-Dimethylhydrazine dihydrochloride, 99+%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
May cause cancer.
Toxic if inhaled.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger