Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sym-Diphenylcarbazone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
10g
25g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

C6H5NHNHCON:NC6H5·C6H5NHNHCONHNHC6H5
sym-Diphenylcarbazone
Baker Analyzed™ ACS Reagent

High quality chemicals for laboratory and specialized industrial use

Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C6H5NHNHCON:NC6H5·C6H5NHNHCONHNHC6H5
Quantity 25g
Formula Weight 482.55
Chemical Name or Material sym-Diphenylcarbazone
Grade Baker Analyzed™ ACS Reagent