Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sym-Diphenylcarbazone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
10g
25g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

538-62-5
C13H12N4O
240.27
ZFWAHZCOKGWUIT-UHFFFAOYSA-N
10860

High quality chemicals for laboratory and specialized industrial use

Packaging Wide Mouth Amber Glass Bottle
Quantity 10g
Formula Weight 482.55
Chemical Name or Material sym-Diphenylcarbazone
Grade Baker Analyzed™ ACS Reagent