Learn More
Waters Corp Symmetry300 C4 Method Validation Kit, 5 μm, 300 Å
Supplier: Waters Corp 186000294
The Symmetry300 C4 Method Validation Kit is designed for the analysis of large peptides and proteins. This kit provides three high-performance C4 columns with 300 Å pore size and 5 μm particle size, ideal for method development and ensuring reproducible results across different batches.
- Suitable for large peptides and proteins
- Features C4 chemistry with 2.8% carbon load
- Pore size of 300 Å
- Particle size of 5 μm
- Column dimensions are 4.6 mm × 150 mm
- Includes three columns from different batches for reproducibility testing
- Provides superior performance in separating biologically active peptides
- Performs well under high and low mass load conditions
- End-capped columns
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.