Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

syn-2-Pyridinealdoxime, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1193-96-0
C6H6N2O
122.13
MFCD00167166
VRGKDCRTYBCJTA-UHFFFAOYSA-N
pyridine-2-aldoxime,picolinaldehyde oxime,2-pyridinecarbaldehyde oxime,z-pyridine-2-carbaldehyde oxime,2-nitrosomethylidene-1h-pyridine,2-pyridinealdoxime,e-picolinaldehyde oxime,2-hydroxyiminomethyl-pyridine,2-pyridine aldoxime,2-pyridinecarboxaldehyde, oxime, e
6740772
2-(nitrosomethylidene)-1H-pyridine
C1=CC(=CN=O)NC=C1
1193-96-0
C6H6N2O
122.13
MFCD00167166
VRGKDCRTYBCJTA-UHFFFAOYSA-N
pyridine-2-aldoxime,picolinaldehyde oxime,2-pyridinecarbaldehyde oxime,z-pyridine-2-carbaldehyde oxime,2-nitrosomethylidene-1h-pyridine,2-pyridinealdoxime,e-picolinaldehyde oxime,2-hydroxyiminomethyl-pyridine,2-pyridine aldoxime,2-pyridinecarboxaldehyde, oxime, e
6740772
2-(nitrosomethylidene)-1H-pyridine
C1=CC(=CN=O)NC=C1
Melting Point 110°C to 113°C
Color Pink to White
Packaging Glass bottle
Quantity 100g
Water 0.2% max. (K.F.)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Beilstein 21, I, 288
Solubility Information Solubility in water: 15g/L (20°C). Other solubilities: may decompose,265g/L ethanol (20°C),31% in methanol- 18% in acetone (20°C),0.5% in toluene- 0.5% in gasoline (20°C)
Formula Weight 122.13
Physical Form Crystalline Powder
Percent Purity 99.0 to 101.0%
Chemical Name or Material syn-2-Pyridinealdoxime
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.