Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syn-7-bromonorbornene, 97%, Acros Organics

Click to view available options
Quantity:
5g
20047-65-8
C7H9Br
173.053
MFCD01862962
XFPAUZYYSFLZRD-MEKDEQNOSA-N
1r,4s-7-bromobicyclo 2.2.1 hept-2-ene,4s,1r-7-bromobicyclo 2.2.1 hept-2-ene,1r,4s,7r-7-bromobicyclo 2.2.1 hept-2-ene,1alpha,4alpha,7-syn-7-bromobicyclo 2.2.1 hepta-2-ene,syn-7-bromobicyclo 2.2.1 hept-2-ene gc
12644178
(1S,4R)-7-bromobicyclo[2.2.1]hept-2-ene
C1CC2C=CC1C2Br
20047-65-8
C7H9Br
173.053
MFCD01862962
XFPAUZYYSFLZRD-MEKDEQNOSA-N
1r,4s-7-bromobicyclo 2.2.1 hept-2-ene,4s,1r-7-bromobicyclo 2.2.1 hept-2-ene,1r,4s,7r-7-bromobicyclo 2.2.1 hept-2-ene,1alpha,4alpha,7-syn-7-bromobicyclo 2.2.1 hepta-2-ene,syn-7-bromobicyclo 2.2.1 hept-2-ene gc
12644178
(1S,4R)-7-bromobicyclo[2.2.1]hept-2-ene
C1CC2C=CC1C2Br
Refractive Index 1.5260 to 1.5300 (20°C, 589nm)
Color Colorless
Density 1.46g/mL
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 97%
Formula Weight 173.05
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material syn-7-Bromonorbornene

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.