Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synapsin I Mouse anti-Human, Mouse, Rat, Clone: 7B1D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261894

Catalog No. NBP2-61894


Add to cart

Description

Description

Synapsin I Monoclonal antibody specifically detects Synapsin I in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Synapsin I
7B1D9
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Purified
SYN1
Purified recombinant fragment of human SYN1 (AA: 362-511) expressed in E. Coli.
74.1 kDa
0.1 ml
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
SYN1a, SYN1b, Synapsin 1, synapsin IBrain protein 4.1, synapsin-1, SYNI
Mouse
IgG1
Protein G purified
Cellular Markers, Circadian Rhythm, Cognition and Behavior, Neuronal Cell Markers, Neuroscience, Neurotransmission, Pain Research, Phospho Specific, Sensory Systems, Vision
Primary
6853
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit