Learn More
Epredia™ Lab Vision™ Synaptophysin Ab-2, Mouse Monoclonal Antibody
Provide accurate, reproducible results with the Epredia™ Synaptophysin Ab-2, Mouse Monoclonal Antibody.
Specifications
Antigen | Synaptophysin Ab-2 |
---|---|
Clone | SYP02 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1150R1
|
Epredia™
MS1150R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
Description
Host Species: Mouse
Clone: SYP02
Isotype: IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: Near C-terminus
Immunogen: A synthetic peptide encoding a region near the C-terminal end of the synaptophysin protein
Molecular Weight: 38kDa
Positive Control: Pancreas, rheochromocytoma or thyroid
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Synaptophysin Ab-2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Rat | |
IgG1 |
SYP02 | |
Monoclonal | |
Mouse | |
Neuroscience | |
A synthetic peptide encoding a region near the C-terminal end of the synaptophysin protein | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.