Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody

Provide accurate, reproducible results Thermo Scientific™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB1461P

 View more versions of this product

Catalog No. RB1461P


Add to cart

Description

Description

Provide accurate, reproducible results Thermo Scientific™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human synaptophysin

Molecular Weight: 38kDa

Positive Control: IMR-5 cells, Pancreas or Pheochromocytoma or GI tumor

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Synaptophysin Ab-4
Polyclonal
1mg/mL
Purified Ab with BSA and Azide
Rabbit
IgG
RUO
Primary
Polyclonal
Western Blot
Polyclonal
Unconjugated
Concentrated
A synthetic peptide from C-terminus of human synaptophysin
1mL
Cancer and Tumor Biology
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit