Learn More
Enzo Life Sciences Synaptophysin monoclonal antibody (EP10) (100 µg)
Supplier: Enzo Life Sciences ADI905782100
Clone: EP10. Host: Mouse. Isotype: IgG1. Immunogen: Immunoprecipitate of human brain. UniProt: P08247. Source: Purified from mouse ascites. Species Reactivity: Human. Applications: WB. Recommended dilutions: Western Blot (1µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.